Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322327.1 | NP_001309256.1 | 4, 5 | 394 | ||
NM_020776.1 | NP_065827.1 | 5, 6 | 478 | |||
NR_136303.1 | 5, 6 | 489 | ||||
NR_136304.1 | 5, 6 | 489 | ||||
NR_136305.1 | 4, 5 | 394 | ||||
XM_005258315.4 | XP_005258372.1 | 5, 6 | 481 | |||
XM_005258317.3 | XP_005258374.1 | 5, 6 | 870 | |||
XM_005258318.4 | XP_005258375.1 | 5, 6 | 485 | |||
XM_011526099.2 | XP_011524401.1 | 5, 6 | 674 | |||
XM_011526100.2 | XP_011524402.1 | 5, 6 | 481 | |||
XM_017025876.1 | XP_016881365.1 | 5, 6 | 791 | |||
XM_017025877.1 | XP_016881366.1 | 5, 6 | 417 | |||
XM_017025878.1 | XP_016881367.1 | 4, 5 | 571 | |||
GenBank mRNA | AB037749.2 | - | 5, 6 | 870 | ||
AK292288.1 | - | 5, 6 | 467 | |||
AK304230.1 | - | 4, 5 | 394 | |||
AL832625.2 | - | 5, 6 | 479 | |||
BX648875.1 | - | 4, 5 | 370 |