Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RD3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257268.1 | NP_001244197.1 | 2 | 447 | ||
GenBank mRNA | BX117646.1 | - | 2 | 447 |