Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005259.2 | NP_005250.1 | 1 | 478 | ||
GenBank mRNA | AF019627.1 | - | 1 | 345 | ||
AF104922.1 | - | 1 | 478 | |||
AK291524.1 | - | 1 | 449 | |||
BC074757.2 | - | 1 | 392 | 30915216 |