Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001354.5 | NP_001345.1 | 6 | 794 | ||
NM_205845.2 | NP_995317.1 | 5 | 694 | |||
GenBank mRNA | AB021654.1 | - | 4 | 381 | ||
AB031084.1 | - | 4 | 381 | |||
AB209017.1 | - | 4 | 448 | |||
AK091194.1 | - | 5 | 2048 | |||
AK290304.1 | - | 4 | 437 | |||
BC007024.1 | - | 4 | 403 | 3681970 | ||
BC063574.1 | - | 5 | 661 | 3877666 | ||
BT006653.1 | - | 4 | 381 | |||
M33376.1 | - | 4 | 336 | |||
S68330.1 | - | 4 | 387 | |||
U05598.1 | - | 4 | 404 |
Gene Symbol: AKR1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001353.5 | NP_001344.2 | 4 | 564 | ||
GenBank mRNA | AB031083.1 | - | 4 | 381 | ||
BC015490.2 | - | 4 | 398 | 3877178 | ||
BC020216.1 | - | 4 | 397 | 4662654 | ||
BC040210.1 | - | 9 | 1223 | 4825338 | ||
BM923712.1 | - | 4 | 413 | |||
BT007197.1 | - | 4 | 381 | |||
D26124.1 | - | 4 | 332 | |||
M86609.1 | - | 4 | 387 | |||
S68290.1 | - | 4 | 372 | |||
U05684.1 | - | 4 | 384 |