Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004536.2 | NP_004527.2 | 17 | 4786 | ||
NM_022892.1 | NP_075043.1 | 15 | 4115 | |||
XM_011543413.2 | XP_011541715.1 | 7 | 2924 | |||
XM_017009490.1 | XP_016864979.1 | 7 | 3471 | |||
GenBank mRNA | AB209123.1 | - | 6 | 4172 | ||
AK124511.1 | - | 6 | 3467 | |||
AK295115.1 | - | 6 | 2014 | |||
BC136273.1 | - | 15 | 4123 | 9020260 | ||
BC143761.1 | - | 14 | 3955 | 9052275 | ||
U19251.1 | - | 17 | 4359 |