Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115156.1 | NP_001108628.1 | 7 | 1019 | ||
NM_001494.3 | NP_001485.2 | 8 | 1154 | |||
XM_017016071.1 | XP_016871560.1 | 8 | 849 | |||
GenBank mRNA | AB451295.1 | - | 8 | 863 | ||
AB451429.1 | - | 8 | 863 | |||
AF144713.1 | - | 5 | 623 | |||
AK094671.1 | - | 1 | 1073 | |||
AK122841.1 | - | 2 | 2327 | |||
AK297177.1 | - | 9 | 1121 | |||
AK297554.1 | - | 7 | 904 | |||
AK309360.1 | - | 9 | 1070 | |||
AK312485.1 | - | 8 | 1039 | |||
BC005145.2 | - | 8 | 947 | 3504736 | ||
BC024168.1 | - | 2 | 92 | |||
BT006868.1 | - | 8 | 863 | |||
BX648038.1 | - | 7 | 700 | |||
CR457073.1 | - | 8 | 863 | |||
D13988.1 | - | 8 | 883 | |||
EU794614.1 | - | 8 | 863 | |||
Y13286.1 | - | 8 | 1015 |