Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034996.2 | NP_001030168.1 | 5, 6 | 497 | ||
NM_003973.4 | NP_003964.3 | 5, 6 | 433 | |||
GenBank mRNA | AK222750.1 | - | 1 | 392 | ||
AK223111.1 | - | 5, 6 | 386 | |||
AK290674.1 | - | 5, 6 | 393 | |||
AK300873.1 | - | 5, 7 | 386 | |||
BC000606.2 | - | 5, 6 | 399 | 3346054 | ||
BC005134.2 | - | 4, 5 | 364 | 3928410 | ||
BC009294.2 | - | 5, 6 | 396 | 4123062 | ||
BC019651.1 | - | 1 | 371 | 4454086 | ||
BC022805.1 | - | 1 | 361 | |||
BC029036.1 | - | 5, 6 | 485 | 5183728 | ||
BC071913.1 | - | 1 | 393 | 6671085 | ||
BG564298.1 | - | 5, 6 | 433 | |||
BG776586.1 | - | 5, 6 | 416 | |||
CD298951.1 | - | 5, 6 | 500 | |||
D87735.1 | - | 5, 6 | 381 | |||
DQ118667.1 | - | 4, 5 | 364 | |||
L10376.1 | - | 1 | 369 | |||
U16738.1 | - | 4, 5 | 370 | |||
U92982.1 | - | 4, 5 | 298 |