Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005262.2 | NP_005253.3 | 3 | 627 | ||
GenBank mRNA | AF124603.1 | - | 2 | 335 | ||
AF124604.1 | - | 3 | 473 | |||
AF208342.1 | - | 3 | 315 | |||
AF306863.1 | - | 3 | 557 | |||
AY550027.1 | - | 3 | 317 | |||
BC002429.1 | - | 3 | 563 | 3346679 | ||
BC028348.2 | - | 3 | 554 | 3636052 | ||
U31176.1 | - | 3 | 468 |