Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127215.1 | NP_001120687.1 | 2 | 241 | ||
NM_001127216.1 | NP_001120688.1 | 2 | 234 | |||
NM_005263.3 | NP_005254.2 | 2 | 327 | |||
XM_005270749.3 | XP_005270806.1 | 2 | 683 | |||
XM_011541245.2 | XP_011539547.1 | 2 | 1169 | |||
XM_011541246.2 | XP_011539548.1 | 2 | 445 | |||
GenBank mRNA | AB209606.1 | - | 1 | 115 | ||
AK123009.1 | - | 2 | 396 | |||
AK313876.1 | - | 2 | 241 | |||
BC032751.1 | - | 2 | 211 | 5585794 | ||
BC074866.2 | - | 1 | 121 | 30915466 | ||
BC074867.2 | - | 1 | 121 | 30915231 | ||
DB129173.1 | - | 2 | 241 | |||
DB137690.1 | - | 2 | 236 | |||
U67369.1 | - | 2 | 344 |