Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004759.4 | NP_004750.1 | 3 | 641 | ||
NM_032960.3 | NP_116584.2 | 3 | 641 | |||
XM_005273353.3 | XP_005273410.1 | 3 | 749 | |||
XM_017002810.1 | XP_016858299.1 | 3 | 307 | |||
GenBank mRNA | AK307499.1 | - | 3 | 226 | ||
BC036060.1 | - | 3 | 641 | 5296259 | ||
BC052584.1 | - | 3 | 624 | 6267183 | ||
X75346.1 | - | 3 | 419 |