Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM227A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013647.1 | NP_001013669.1 | 10 | 1537 | ||
NM_001291030.1 | NP_001277959.1 | 10 | 1577 | |||
XM_006724291.2 | XP_006724354.2 | 10 | 1537 | |||
XM_006724292.2 | XP_006724355.2 | 10 | 1537 | |||
XM_006724293.2 | XP_006724356.1 | 10 | 1537 | |||
XM_006724294.2 | XP_006724357.2 | 10 | 1577 | |||
XM_006724295.2 | XP_006724358.2 | 10 | 1577 | |||
XM_011530308.2 | XP_011528610.1 | 10 | 1242 | |||
XM_011530309.2 | XP_011528611.1 | 10 | 1537 | |||
XM_011530310.1 | XP_011528612.1 | 9 | 1432 | |||
XM_011530311.1 | XP_011528613.1 | 9 | 1414 | |||
XM_011530312.1 | XP_011528614.1 | 8 | 814 | |||
XM_011530313.2 | XP_011528615.1 | 8 | 766 | |||
XM_011530314.1 | XP_011528616.1 | 9 | 1467 | |||
GenBank mRNA | AK301819.1 | - | 10 | 1251 | ||
DV080205.1 | - | 2 | 101 |