Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000821.6 | NP_000812.2 | 13 | 1872 | ||
NM_001142269.3 | NP_001135741.1 | 12 | 1701 | |||
XM_005264259.4 | XP_005264316.1 | 13 | 1831 | |||
XM_011532764.2 | XP_011531066.1 | 12 | 1667 | |||
XM_011532765.2 | XP_011531067.1 | 11 | 1506 | |||
XM_017003803.1 | XP_016859292.1 | 12 | 1694 | |||
GenBank mRNA | AK130139.1 | - | 4 | 464 | ||
AK222827.1 | - | 13 | 1832 | |||
AK225663.1 | - | 13 | 1831 | |||
AK297397.1 | - | 12 | 1641 | |||
AK298982.1 | - | 12 | 1526 | |||
AK313126.1 | - | 13 | 1840 | |||
BC013979.2 | - | 13 | 1794 | 4129593 | ||
EU847509.1 | - | 13 | 1769 | |||
L17128.1 | - | 13 | 1921 | |||
M81592.1 | - | 13 | 1852 |