Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AATK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080395.2 | NP_001073864.2 | 11 | 1672 | ||
NM_004920.2 | NP_004911.2 | 10 | 1450 | |||
XM_006722193.2 | XP_006722256.1 | 11 | 1665 | |||
XM_006722194.2 | XP_006722257.1 | 10 | 1517 | |||
XM_006722195.2 | XP_006722258.1 | 9 | 1459 | |||
XM_006722196.2 | XP_006722259.1 | 11 | 1634 | |||
XM_011525509.2 | XP_011523811.1 | 10 | 3101 | |||
XM_011525510.2 | XP_011523812.1 | 10 | 1477 | |||
XM_011525511.2 | XP_011523813.1 | 6 | 1330 | |||
XM_017025421.1 | XP_016880910.1 | 10 | 1464 | |||
XM_017025422.1 | XP_016880911.1 | 10 | 1623 | |||
GenBank mRNA | AB014541.2 | - | 10 | 1450 | ||
AK131395.1 | - | 10 | 1354 | |||
AK131529.1 | - | 12 | 1564 |