Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001497.3 | NP_001488.2 | 4 | 1060 | ||
GenBank mRNA | A23697.1 | - | 4 | 877 | ||
AK296928.1 | - | 5 | 759 | |||
AK297541.1 | - | 3 | 394 | |||
AK312797.1 | - | 4 | 899 | |||
AK316294.1 | - | 5 | 732 | |||
BX648438.1 | - | 4 | 318 | |||
M13701.1 | - | 3 | 460 | |||
M22921.1 | - | 4 | 943 | |||
U10472.1 | - | 2 | 96 | |||
U10473.1 | - | 4 | 601 | |||
U10474.1 | - | 4 | 944 | |||
X13223.1 | - | 4 | 943 | |||
X14085.1 | - | 4 | 940 | |||
X55415.1 | - | 4 | 871 |