Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178026.2 | NP_821158.2 | 4 | 621 | ||
XM_011528782.2 | XP_011527084.1 | 2 | 58 | |||
XM_017027809.1 | XP_016883298.1 | 4 | 620 | |||
XM_017027810.1 | XP_016883299.1 | 4 | 620 | |||
XM_017027811.1 | XP_016883300.1 | 4 | 620 | |||
GenBank mRNA | AK097100.1 | - | 3 | 479 | ||
AK124788.1 | - | 3 | 935 | |||
AK299494.1 | - | 4 | 619 | |||
AY138815.1 | - | 4 | 578 | |||
BC033745.1 | - | 4 | 621 | |||
BC040030.1 | - | 5 | 636 | |||
BC101371.1 | - | 4 | 610 | |||
BC101372.1 | - | 4 | 617 | |||
BX401449.2 | - | 4 | 628 | |||
CA454346.1 | - | 4 | 617 |