Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290224.1 | NP_001277153.1 | 18 | 1587 | ||
NM_199259.3 | NP_954868.1 | 22 | 1809 | |||
NM_199260.3 | NP_954869.1 | 21 | 1749 | |||
NM_199261.3 | NP_954870.2 | 23 | 1863 | |||
GenBank mRNA | AF007118.1 | - | 23 | 1811 | ||
AF495908.1 | - | 22 | 1757 | |||
AK308254.1 | - | 22 | 1706 | |||
AK314285.1 | - | 23 | 1762 | |||
AL833345.1 | - | 23 | 1770 | |||
AY219886.1 | - | 22 | 1757 | |||
AY219887.1 | - | 21 | 1697 | |||
AY219888.1 | - | 18 | 1535 | |||
BC028719.1 | - | 22 | 1792 | 5269598 |