Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002059.4 | NP_002050.1 | 3 | 418 | ||
NM_022556.3 | NP_072050.1 | 3 | 373 | |||
NM_022557.3 | NP_072051.1 | 3 | 418 | |||
NM_022558.3 | NP_072052.1 | 3 | 418 | |||
GenBank mRNA | AB451332.1 | - | 2 | 232 | ||
AB451477.1 | - | 2 | 232 | |||
AF006060.1 | - | 3 | 244 | |||
AF006061.1 | - | 2 | 277 | |||
AK312194.1 | - | 3 | 340 | |||
BC020760.1 | - | 3 | 339 | 4720475 | ||
CR541924.1 | - | 2 | 277 | |||
J03756.1 | - | 3 | 338 | |||
M38451.1 | - | 3 | 289 |