Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003090.3 | NP_003081.2 | 5 | 496 | ||
NR_135506.1 | 3 | 269 | ||||
NR_135507.1 | 2 | 222 | ||||
NR_135508.1 | 5 | 496 | ||||
GenBank mRNA | AJ130971.1 | - | 2 | 123 | ||
AJ130972.1 | - | 3 | 170 | |||
AK090986.1 | - | 2 | 830 | |||
AK223070.1 | - | 5 | 453 | |||
AK223110.1 | - | 5 | 418 | |||
AK290078.1 | - | 5 | 452 | |||
AK312200.1 | - | 5 | 427 | |||
AL538391.3 | - | 5 | 495 | |||
BC022816.1 | - | 5 | 449 | 4904205 | ||
BC067846.1 | - | 5 | 465 | 30330402 | ||
BC071717.1 | - | 1 | 404 | 4514975 | ||
X13482.1 | - | 5 | 448 |