Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301206.1 | NP_001288135.1 | 5 | 3110 | ||
NM_001717.3 | NP_001708.3 | 5 | 2974 | |||
XM_011521893.1 | XP_011520195.1 | 5 | 2876 | |||
XM_011521894.1 | XP_011520196.1 | 4 | 2792 | |||
GenBank mRNA | AK302992.1 | - | 6 | 2990 | ||
L03427.1 | - | 2970 |