Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000823.3 | NP_000814.2 | 5 | 499 | ||
XM_011515263.1 | XP_011513565.1 | 2 | 517 | |||
GenBank mRNA | AF282259.1 | - | 2 | 517 | ||
AF282260.1 | - | 2 | 517 | |||
AF282261.1 | - | 3 | 341 | |||
AY557192.1 | - | 5 | 451 | |||
L01406.1 | - | 5 | 502 | |||
L09237.1 | - | 5 | 499 |
Set Membership: |
G-protein coupled receptor |