Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NAV2 SIRNA Assays ›
Gene Symbol:
NAV2
Gene Name:
neuron navigator 2
Gene Aliases:
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location:
Chr.11: 19345200 - 20121601 on Build GRCh38
UniGene ID:
Hs.502116
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s57499
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NAV2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001111018.1
NP_001104488.1
7
1994
NM_001244963.1
NP_001231892.1
8
2387
NM_145117.4
NP_660093.2
7
2318
NM_182964.5
NP_892009.3
7
2318
XM_006718364.3
XP_006718427.1
7
2640
XM_006718365.3
XP_006718428.1
8
2709
XM_006718366.3
XP_006718429.1
8
1873
XM_011520444.2
XP_011518746.1
8
2709
XM_011520445.2
XP_011518747.1
8
2709
XM_011520446.2
XP_011518748.1
8
2709
XM_011520447.2
XP_011518749.1
8
2709
XM_011520448.2
XP_011518750.1
8
2580
XM_011520449.2
XP_011518751.1
8
2709
XM_011520450.2
XP_011518752.1
8
2709
XM_011520451.2
XP_011518753.1
7
2087
XM_011520452.2
XP_011518754.1
8
2457
XM_017018520.1
XP_016874009.1
9
2501
XM_017018521.1
XP_016874010.1
8
2709
XM_017018522.1
XP_016874011.1
8
2457
XM_017018523.1
XP_016874012.1
8
3386
XM_017018524.1
XP_016874013.1
8
2209
XM_017018525.1
XP_016874014.1
2
1052
GenBank mRNA
AB037840.3
-
7
2319
AB063115.1
-
7
1994
AB063116.1
-
7
2287
AF466143.1
-
7
2033
AF466144.1
-
7
2033
AJ488102.1
-
8
2125
BC150290.1
-
7
2319
8860130
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure by baroreceptor feedback
sensory perception of sound
sensory perception of smell
locomotory behavior
optic nerve development
glossopharyngeal nerve development
vagus nerve development
helicase activity
protein binding
ATP binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline