Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256196.1 | NP_001243125.1 | 12 | 1510 | ||
NM_001654.4 | NP_001645.1 | 12 | 1501 | |||
XM_006724529.2 | XP_006724592.1 | 12 | 1437 | |||
XM_011543906.2 | XP_011542208.1 | 12 | 2827 | |||
XM_011543908.2 | XP_011542210.1 | 12 | 2811 | |||
XM_011543909.2 | XP_011542211.1 | 7 | 802 | |||
GenBank mRNA | AB208831.1 | - | 9 | 4063 | ||
AF052136.1 | - | 2 | 208 | |||
AK130043.1 | - | 12 | 1394 | |||
AK300975.1 | - | 12 | 1357 | |||
BC002466.2 | - | 12 | 1381 | 3347509 | ||
BC007514.2 | - | 12 | 1388 | 2966604 | ||
BT019864.1 | - | 11 | 1290 | |||
KU177914.1 | - | 11 | 1290 | |||
KU177915.1 | - | 11 | 1305 | |||
M13829.1 | - | 3 | 417 | |||
X04790.1 | - | 12 | 1484 |