Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD99P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033381.1 | 10 | 821 | |||
GenBank mRNA | AK023533.1 | - | 6 | 2459 | ||
BC019893.1 | - | 10 | 692 |