Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166347.1 | NP_001159819.1 | 5, 6 | 814 | ||
NM_001166348.1 | NP_001159820.1 | 4, 5 | 744 | |||
NM_001166349.1 | NP_001159821.1 | 4, 5 | 614 | |||
NM_173626.3 | NP_775897.3 | 5, 6 | 811 | |||
XM_006721833.1 | XP_006721896.1 | 5, 6 | 886 | |||
XM_011524652.1 | XP_011522954.1 | 4, 5 | 790 | |||
XM_011524653.1 | XP_011522955.1 | 4, 5 | 718 | |||
XM_011524654.1 | XP_011522956.1 | 4, 5 | 603 | |||
XM_017024505.1 | XP_016879994.1 | 4, 5 | 800 | |||
XM_017024506.1 | XP_016879995.1 | 4, 5 | 800 | |||
XM_017024507.1 | XP_016879996.1 | 5, 6 | 860 | |||
GenBank mRNA | AF345195.1 | - | 4, 5 | 546 | ||
AJ544073.1 | - | 5, 6 | 785 | |||
AK075248.1 | - | 5, 6 | 774 | |||
AK315132.1 | - | 5, 6 | 777 | |||
AL833468.1 | - | 4, 5 | 708 | |||
BC035900.1 | - | 5, 6 | 753 | 5241643 |