Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM49C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195234.1 | NP_001182163.1 | 3 | 512 | ||
XM_017018126.1 | XP_016873615.1 | 1 | 187 | |||
GenBank mRNA | BC126470.1 | - | 2 | 306 | 8992186 |
Gene Symbol: TRIM49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075019.2 | - | 2 | 211 | 30915409 | |
BC075020.2 | - | 2 | 211 | 30915620 |