Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 11 | 1128 | ||
NM_001278923.1 | NP_001265852.1 | 9 | 957 | |||
NM_001278924.1 | NP_001265853.1 | 8 | 975 | |||
GenBank mRNA | AK223149.1 | - | 11 | 942 | ||
AK300191.1 | - | 9 | 791 | |||
AK313626.1 | - | 11 | 991 | |||
BC008813.2 | - | 8 | 889 | 3958020 | ||
X16662.1 | - | 11 | 972 |
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040084.2 | NP_001035173.1 | 11 | 1091 | ||
NM_001271702.1 | NP_001258631.1 | 11 | 1205 | |||
NM_001271703.1 | NP_001258632.1 | 9 | 905 | |||
XM_006717951.3 | XP_006718014.1 | 11 | 1045 | |||
XM_011540096.1 | XP_011538398.1 | 12 | 1244 | |||
XM_011540098.1 | XP_011538400.1 | 11 | 1148 | |||
GenBank mRNA | AK296573.1 | - | 9 | 756 | ||
AK298761.1 | - | 11 | 1071 | |||
AK300139.1 | - | 11 | 1056 | |||
AK300158.1 | - | 10 | 869 | |||
BC004376.1 | - | 11 | 988 | 3639064 | ||
BC073755.1 | - | 11 | 1022 | 6538697 | ||
M81844.1 | - | 11 | 893 |