Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100134822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL137655.1 | - | 17 | 3246 |
Gene Symbol: PCMTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC032332.1 | - | 7 | 1305 | 5169056 |