Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256196.1 | NP_001243125.1 | 5 | 650 | ||
NM_001256197.1 | NP_001243126.1 | 5 | 650 | |||
NM_001654.4 | NP_001645.1 | 5 | 650 | |||
XM_006724529.2 | XP_006724592.1 | 5 | 571 | |||
XM_011543906.2 | XP_011542208.1 | 5 | 1961 | |||
XM_011543908.2 | XP_011542210.1 | 5 | 1960 | |||
GenBank mRNA | AB158254.1 | - | 4 | 447 | ||
AB208831.1 | - | 5 | 547 | |||
AK130043.1 | - | 5 | 543 | |||
AK297453.1 | - | 5 | 543 | |||
AK307747.1 | - | 5 | 543 | |||
BC002466.2 | - | 5 | 530 | 3347509 | ||
BC007514.2 | - | 5 | 528 | 2966604 | ||
BT019864.1 | - | 4 | 439 | |||
KU177914.1 | - | 4 | 439 | |||
KU177915.1 | - | 4 | 439 | |||
X04790.1 | - | 5 | 633 |