Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTF3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136497.2 | NP_001129969.1 | 3 | 465 | ||
NM_152265.4 | NP_689478.1 | 4 | 532 | |||
GenBank mRNA | AK027582.1 | - | 3 | 346 | ||
AK027750.1 | - | 4 | 414 | |||
BC021004.1 | - | 3 | 309 | |||
BC022371.1 | - | 4 | 394 | 4739327 | ||
BC070378.1 | - | 4 | 422 | 6495112 | ||
BU567198.1 | - | 4 | 535 |