Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130440.1 | NP_001123912.1 | 4 | 459 | ||
NM_003133.5 | NP_003124.1 | 3 | 335 | |||
GenBank mRNA | AF070649.1 | - | 3 | 314 | ||
AK289595.1 | - | 3 | 300 | |||
AL050363.1 | - | 1 | 264 | |||
BC008443.1 | - | 3 | 319 | 4102222 | ||
BC015094.1 | - | 1 | 306 | 3930047 | ||
BC017414.1 | - | 1 | 86 | |||
BC021995.1 | - | 4 | 439 | 4101294 | ||
BC022415.1 | - | 1 | 301 | 4243562 | ||
BC064351.1 | - | 3 | 1551 | 4452218 | ||
BC066957.1 | - | 3 | 322 | 5263635 | ||
BC067845.1 | - | 4 | 394 | 5287027 | ||
BC106005.1 | - | 1 | 292 | 6026221 | ||
DA983529.1 | - | 3 | 335 | |||
DB194289.1 | - | 4 | 464 | |||
EF488978.1 | - | 4 | 443 | |||
U20998.1 | - | 3 | 313 |