Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP4-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033059.3 | NP_149048.2 | 1 | 719 | ||
GenBank mRNA | AJ406944.1 | - | 1 | 725 | ||
BC126131.1 | - | 1 | 701 | 8991847 | ||
BC130562.1 | - | 1 | 701 | 40146593 |
Gene Symbol: KRTAP4-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322457.1 | NP_001309386.1 | 1 | 720 | ||
GenBank mRNA | AJ406940.1 | - | 1 | 437 |