Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079516.3 | NP_001072984.1 | 6 | 887 | ||
NM_001321957.1 | NP_001308886.1 | 6 | 868 | |||
NM_001321958.1 | NP_001308887.1 | 6 | 942 | |||
NM_001321959.1 | NP_001308888.1 | 6 | 961 | |||
NM_001321960.1 | NP_001308889.1 | 7 | 1030 | |||
NR_135904.1 | 5 | 795 | ||||
GenBank mRNA | AK294369.1 | - | 6 | 694 | ||
CN428611.1 | - | 5 | 537 |
Gene Symbol: ASAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143974.1 | NP_001137446.1 | 19 | 2302 | ||
NM_019893.2 | NP_063946.2 | 20 | 2407 | |||
XM_011539970.2 | XP_011538272.1 | 21 | 2603 | |||
XM_011539971.2 | XP_011538273.1 | 20 | 2460 | |||
XM_011539972.2 | XP_011538274.1 | 20 | 2427 | |||
XM_017016435.1 | XP_016871924.1 | 22 | 2574 | |||
XM_017016436.1 | XP_016871925.1 | 6 | 666 | |||
GenBank mRNA | BG289059.1 | - | 4 | 459 |