Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079516.3 | NP_001072984.1 | 6 | 927 | ||
NM_001321957.1 | NP_001308886.1 | 6 | 908 | |||
NM_001321958.1 | NP_001308887.1 | 6 | 982 | |||
NM_001321959.1 | NP_001308888.1 | 6 | 1001 | |||
NM_001321960.1 | NP_001308889.1 | 7 | 1070 | |||
NR_135904.1 | 5 | 835 | ||||
GenBank mRNA | AK294369.1 | - | 6 | 734 |
Gene Symbol: ASAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539970.2 | XP_011538272.1 | 21 | 2643 | ||
XM_011539971.2 | XP_011538273.1 | 20 | 2500 | |||
XM_011539972.2 | XP_011538274.1 | 20 | 2467 | |||
XM_017016435.1 | XP_016871924.1 | 22 | 2614 | |||
XM_017016436.1 | XP_016871925.1 | 6 | 706 |