Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022148.3 | NP_071431.2 | 2 | 219 | ||
NR_110830.1 | 2 | 216 | ||||
XM_011546181.2 | XP_011544483.1 | 2 | 254 | |||
XM_011546182.2 | XP_011544484.1 | 2 | 258 | |||
GenBank mRNA | AB052639.1 | - | 2 | 130 | ||
AF142570.1 | - | 2 | 146 | |||
AF338733.1 | - | 2 | 130 | |||
AK026800.1 | - | 2 | 218 | |||
AK223303.1 | - | 2 | 246 | |||
AK225583.1 | - | 2 | 218 |