Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005267.4 | NP_005258.2 | 2 | 465 | ||
XM_011509416.1 | XP_011507718.1 | 1 | 1212 | |||
XM_011509417.1 | XP_011507719.1 | 1 | 1212 | |||
GenBank mRNA | CD674975.1 | - | 2 | 465 | ||
DB476698.1 | - | 1 | 102 | |||
KJ534845.1 | - | 1 | 412 |