Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM183B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079809.1 | NP_001073277.1 | 1 | 994 | ||
GenBank mRNA | AK297222.1 | - | 7 | 914 |
Gene Symbol: TMEM183A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322955.1 | NP_001309884.1 | 7 | 1024 | ||
NM_001322956.1 | NP_001309885.1 | 7 | 1032 | |||
NM_001322957.1 | NP_001309886.1 | 7 | 1027 | |||
NM_001322958.1 | NP_001309887.1 | 6 | 945 | |||
NM_001322959.1 | NP_001309888.1 | 6 | 942 | |||
NM_138391.4 | NP_612400.3 | 7 | 996 | |||
NR_136530.1 | 8 | 1199 | ||||
NR_136531.1 | 7 | 1012 | ||||
NR_136532.1 | 7 | 1015 | ||||
NR_136533.1 | 8 | 1196 | ||||
NR_136534.1 | 8 | 1196 | ||||
GenBank mRNA | AF070537.1 | - | 7 | 1013 | ||
AK291426.1 | - | 7 | 1010 | |||
AK312985.1 | - | 7 | 1010 | |||
BC013073.1 | - | 1 | 996 | 4151716 | ||
BC038952.2 | - | 1 | 1048 | 5740484 | ||
BC067775.1 | - | 8 | 1170 | 4817183 | ||
BC150647.1 | - | 7 | 978 | 9057018 |