Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286401.1 | NP_001273330.1 | 3 | 1355 | ||
NM_145316.3 | NP_660359.2 | 4 | 922 | |||
NR_104442.1 | 3 | 824 | ||||
NR_104443.1 | 2 | 739 | ||||
XM_011514367.2 | XP_011512669.1 | 4 | 849 | |||
XM_011514369.2 | XP_011512671.1 | 2 | 250 | |||
XM_011514370.2 | XP_011512672.1 | 2 | 240 | |||
GenBank mRNA | AK098666.1 | - | 4 | 881 | ||
BC026012.1 | - | 3 | 794 | 5269554 | ||
BC029657.1 | - | 2 | 220 | 5167418 | ||
DB458508.1 | - | 3 | 306 |