Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263741.4 | XP_005263798.1 | 3 | 1626 | ||
GenBank mRNA | AK090531.1 | - | 1 | 2165 |