Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097622.1 | NP_001091091.1 | 2, 3 | 282 | ||
XM_011515485.2 | XP_011513787.1 | 3, 4 | 229 | |||
GenBank mRNA | BC069468.1 | - | 2, 3 | 190 | 7262729 | |
BC069512.1 | - | 2, 3 | 190 | 7262741 | ||
BC069550.1 | - | 2, 3 | 190 | 7262753 | ||
BC113704.1 | - | 2, 3 | 282 | 8322756 | ||
BC113706.1 | - | 2, 3 | 282 | 8322758 | ||
BC147028.1 | - | 2, 3 | 284 | 9056956 | ||
BC147029.1 | - | 2, 3 | 284 | 9056957 |
Gene Symbol: OCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006188.3 | NP_006179.2 | 2, 3 | 190 |