Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039029.2 | NP_001034118.1 | 5 | 1837 | ||
NM_001163925.1 | NP_001157397.1 | 5 | 1873 | |||
NM_001163926.1 | NP_001157398.1 | 5 | 1840 | |||
XM_011521015.1 | XP_011519317.1 | 5 | 2058 | |||
XM_017019848.1 | XP_016875337.1 | 3 | 1245 | |||
GenBank mRNA | AK095610.1 | - | 5 | 1575 | ||
AK124687.1 | - | 5 | 1444 | |||
AK125402.1 | - | 2 | 661 | |||
AK126866.1 | - | 1 | 1865 | |||
BC036868.1 | - | 1 | 294 | |||
BC146894.1 | - | 5 | 1476 | 9056822 | ||
BC146910.1 | - | 5 | 1476 | 9056838 | ||
BC150568.1 | - | 3 | 1081 | 40127648 |