Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039029.2 | NP_001034118.1 | 5 | 1573 | ||
NM_001163925.1 | NP_001157397.1 | 5 | 1609 | |||
NM_001163926.1 | NP_001157398.1 | 5 | 1576 | |||
XM_011521015.1 | XP_011519317.1 | 5 | 1794 | |||
XM_017019848.1 | XP_016875337.1 | 3 | 981 | |||
GenBank mRNA | AK095610.1 | - | 5 | 1311 | ||
AK124687.1 | - | 5 | 1180 | |||
AK125402.1 | - | 2 | 414 | |||
AK126866.1 | - | 1 | 1601 | |||
BC036868.1 | - | 1 | 30 | |||
BC146894.1 | - | 5 | 1212 | 9056822 | ||
BC146910.1 | - | 5 | 1212 | 9056838 | ||
BC150568.1 | - | 3 | 817 | 40127648 |