Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCP4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102566.1 | NP_001096036.1 | 3 | 997 | ||
XM_017002154.1 | XP_016857643.1 | 3 | 971 | |||
GenBank mRNA | BC028905.2 | - | 3 | 998 | ||
BC067884.1 | - | 3 | 975 | |||
BC147003.1 | - | 3 | 898 | 9056931 | ||
BC147004.1 | - | 3 | 898 | 9056932 | ||
CD237602.1 | - | 1 | 672 |