Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000579.3 | NP_000570.1 | 3 | 857 | ||
NM_001100168.1 | NP_001093638.1 | 3 | 622 | |||
GenBank mRNA | AF011500.1 | - | 1 | 504 | ||
AF011501.1 | - | 1 | 504 | |||
AF011502.1 | - | 1 | 504 | |||
AF011503.1 | - | 1 | 504 | |||
AF011504.1 | - | 1 | 503 | |||
AF011505.1 | - | 1 | 504 | |||
AF011506.1 | - | 1 | 504 | |||
AF011507.1 | - | 1 | 504 | |||
AF011508.1 | - | 1 | 504 | |||
AF011509.1 | - | 1 | 504 | |||
AF011510.1 | - | 1 | 504 | |||
AF011511.1 | - | 1 | 504 | |||
AF011512.1 | - | 1 | 504 | |||
AF011513.1 | - | 1 | 504 | |||
AF011514.1 | - | 1 | 504 | |||
AF011515.1 | - | 1 | 504 | |||
AF011516.1 | - | 1 | 504 | |||
AF011517.1 | - | 1 | 504 | |||
AF011518.1 | - | 1 | 504 | |||
AF011519.1 | - | 1 | 504 | |||
AF011520.1 | - | 1 | 504 | |||
AF011521.1 | - | 1 | 504 | |||
AF011522.1 | - | 1 | 504 | |||
AF011523.1 | - | 1 | 504 | |||
AF011524.1 | - | 1 | 504 | |||
AF011525.1 | - | 1 | 504 | |||
AF011526.1 | - | 1 | 504 | |||
AF011527.1 | - | 1 | 504 | |||
AF011528.1 | - | 1 | 504 | |||
AF011529.1 | - | 1 | 504 | |||
AF011530.1 | - | 1 | 504 | |||
AF011531.1 | - | 1 | 504 | |||
AF011532.1 | - | 1 | 504 | |||
AF011533.1 | - | 1 | 504 | |||
AF011534.1 | - | 1 | 504 | |||
AF011535.1 | - | 1 | 504 | |||
AF011536.1 | - | 1 | 504 | |||
AF011537.1 | - | 1 | 504 | |||
AK292132.1 | - | 2 | 554 | |||
AK314882.1 | - | 2 | 646 | |||
BC038398.1 | - | 2 | 543 | 5186388 | ||
U54994.1 | - | 2 | 554 | |||
U57840.1 | - | 2 | 526 |