Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000404.3 | NP_000395.2 | 3 | 496 | ||
NM_001079811.2 | NP_001073279.1 | 3 | 390 | |||
NM_001317040.1 | NP_001303969.1 | 4 | 640 | |||
GenBank mRNA | AK130133.1 | - | 3 | 424 | ||
AK222763.1 | - | 3 | 387 | |||
AK223091.1 | - | 3 | 412 | |||
AK300021.1 | - | 3 | 323 | |||
AK300753.1 | - | 4 | 556 | |||
AK312988.1 | - | 3 | 383 | |||
AU134179.1 | - | 3 | 468 | |||
BC007493.2 | - | 3 | 398 | 2988086 | ||
BT007147.1 | - | 3 | 351 | |||
BX485230.1 | - | 3 | 418 | |||
DA077250.1 | - | 3 | 412 | |||
DA261513.1 | - | 4 | 502 | |||
DB504658.1 | - | 3 | 380 | |||
DC301575.1 | - | 3 | 496 | |||
M22590.1 | - | 3 | 385 | |||
M27507.1 | - | 3 | 401 | |||
M34423.1 | - | 3 | 411 |