Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083124.1 | NP_001076593.1 | 4 | 3369 | ||
XM_011545680.2 | XP_011543982.1 | 3 | 3286 | |||
GenBank mRNA | AL080148.1 | - | 1 | 2324 |