Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308253.1 | NP_001295182.1 | 6 | 1130 | ||
NM_002061.3 | NP_002052.1 | 7 | 1196 | |||
XM_011541261.2 | XP_011539563.1 | 7 | 997 | |||
XM_017001056.1 | XP_016856545.1 | 8 | 1398 | |||
XM_017001057.1 | XP_016856546.1 | 7 | 1332 | |||
GenBank mRNA | AB809606.1 | - | 6 | 785 | ||
AK290449.1 | - | 7 | 1195 | |||
BC041809.1 | - | 7 | 997 | 5311598 | ||
CR541833.1 | - | 7 | 761 | |||
L35546.1 | - | 7 | 1014 |