Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040094.1 | 5 | 736 | |||
GenBank mRNA | BC047459.1 | - | 5 | 736 |
Gene Symbol: LOC101927079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110480.1 | 3 | 249 | |||
GenBank mRNA | AK058056.1 | - | 3 | 249 | ||
DB049942.1 | - | 3 | 249 |
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546205.1 | XP_011544507.1 | 3 | 249 | ||
XM_011546206.1 | XP_011544508.1 | 3 | 249 |
Gene Symbol: LOC727924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015416.1 | 3 | 248 | |||
GenBank mRNA | BC048276.1 | - | 3 | 248 |