Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032498.2 | NP_115887.1 | 3 | 700 | ||
XM_005262484.3 | XP_005262541.1 | 3 | 714 | |||
GenBank mRNA | AF317219.1 | - | 3 | 640 | ||
AK058125.1 | - | 3 | 593 | |||
AL590526.1 | - | 3 | 700 | |||
BC021719.2 | - | 3 | 595 | 5269333 |
Gene Symbol: RHOXF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099685.1 | NP_001093155.1 | 3 | 700 | ||
GenBank mRNA | AL590524.1 | - | 3 | 596 |