Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003722.1 | NP_001003722.1 | 11 | 1700 | ||
NM_001499.2 | NP_001490.1 | 11 | 1700 | |||
XM_006717059.2 | XP_006717122.1 | 13 | 1839 | |||
XM_006717060.2 | XP_006717123.1 | 12 | 1812 | |||
XM_011518549.1 | XP_011516851.1 | 13 | 1876 | |||
XM_011518550.1 | XP_011516852.1 | 13 | 1727 | |||
XM_011518551.1 | XP_011516853.1 | 12 | 1727 | |||
GenBank mRNA | AF058922.1 | - | 11 | 1673 | ||
AK001767.1 | - | 6 | 950 | |||
AK074801.1 | - | 10 | 1456 | |||
AK222842.1 | - | 11 | 1646 | |||
AK222854.1 | - | 11 | 1674 | |||
AK290533.1 | - | 10 | 1437 | |||
AL117584.1 | - | 9 | 1183 | |||
BC030012.1 | - | 11 | 1693 | 4829102 |