Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015201.4 | NP_056016.1 | 8, 9 | 1207 | ||
GenBank mRNA | AB060694.1 | - | 7, 8 | 875 | ||
AK024840.1 | - | 8, 9 | 1180 | |||
AK290677.1 | - | 8, 9 | 1169 | |||
AK293719.1 | - | 7, 8 | 851 | |||
AK310015.1 | - | 6, 7 | 781 | |||
AK315612.1 | - | 8, 9 | 1176 | |||
BC001086.2 | - | 7, 8 | 933 | |||
BC005160.2 | - | 7, 8 | 960 | |||
BC007274.1 | - | 8, 9 | 1102 | |||
BC013787.1 | - | 8, 9 | 1169 | 3872212 | ||
BC013980.2 | - | 8, 9 | 1172 | 3633864 | ||
BC017674.2 | - | 8, 9 | 1159 | 3905068 | ||
BC073813.1 | - | 3, 4 | 466 | |||
BC098446.1 | - | 4, 5 | 603 | |||
D50914.1 | - | 7, 8 | 947 |